Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Oxaspiro[4.4]nonan-3-ol
1-Oxaspiro[4.4]nonan-3-ol
1-Oxaspiro[4.4]nonan-3-ol
1-Oxaspiro[4.4]nonan-3-ol
Supplier:
Apollo Scientific
Certificates
Expand image
1-Oxaspiro[4.4]nonan-3-ol
Supplier:
Apollo Scientific
About this item
CAS Number:
1331825-49-0
:
Formula:
C₈H₁₄O₂
MDL Number:
MFCD30471614
:
142.196 g/mol
Storage Temperature:
Ambient