Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Azaspiro[2.3]hexane hydrochloride
5-Azaspiro[2.3]hexane hydrochloride
5-Azaspiro[2.3]hexane hydrochloride
5-Azaspiro[2.3]hexane hydrochloride
Supplier:
Apollo Scientific
Certificates
Expand image
5-Azaspiro[2.3]hexane hydrochloride
Supplier:
Apollo Scientific
About this item
CAS Number:
1536169-63-7
:
Formula:
C₅H₉N·ClH
MDL Number:
MFCD27991827
:
119.593 g/mol
Storage Temperature:
Refrigerator