6-Oxa-1-azaspiro[3.5]nonane
Catalog # APOSOR304426-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:127.184 g/mol
Storage Temperature:Ambient
CAS Number:1214875-46-3
Formula:C₇H₁₃NO
MDL Number:MFCD14581253