2,7-Diazaspiro[3.5]nonane dihydrochloride
Catalog # APOSOR304422-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:199.121 g/mol
Storage Temperature:Ambient
CAS Number:1334499-85-2
Formula:C₇H₁₄N₂·2ClH
MDL Number:MFCD20261446