Order Entry
Export
ContactUsLinkComponent
Spiro[3.5]nonan-1-one
Spiro[3.5]nonan-1-one
Catalog # APOSOR304419-1G
Supplier:  Apollo Scientific
CAS Number:  
Spiro[3.5]nonan-1-one
Catalog # APOSOR304419-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :138.207 g/mol
Storage Temperature:Ambient
CAS Number:29800-45-1
Formula:C₉H₁₄O
MDL Number:MFCD24386783