Spiro[3.5]nonan-1-ol
Catalog # APOSOR304418-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:140.223 g/mol
Storage Temperature:Ambient
CAS Number:60211-19-0
Formula:C₉H₁₆O
MDL Number:MFCD26407277