Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Oxaspiro[3.5]nonan-8-one
5-Oxaspiro[3.5]nonan-8-one
5-Oxaspiro[3.5]nonan-8-one
APOSOR304416-1G
:
Apollo Scientific
:
5-Oxaspiro[3.5]nonan-8-one
APOSOR304416-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
140.18 g/mol
:
Ambient
:
1367945-40-1
:
C₈H₁₂O₂
:
MFCD22070990