1,6-Diazaspiro[3.5]nonan-2-one
Catalog # APOSOR304414-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:140.183 g/mol
Storage Temperature:Ambient
CAS Number:1567085-15-7
Formula:C₇H₁₂N₂O
MDL Number:MFCD26858689