Order Entry
Export
ContactUsLinkComponent
 
7-Methyl-2,7-diazaspiro[4.5]decane dihydrochloride
Catalog # APOSOR304410-1G
Supplier:  Apollo Scientific
CAS Number:  
7-Methyl-2,7-diazaspiro[4.5]decane dihydrochloride
Catalog # APOSOR304410-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :227.175 g/mol
Storage Temperature:Ambient
CAS Number:1086395-12-1
Formula:C₉H₁₈N₂·2ClH
MDL Number:MFCD26407167