Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,9-Diazaspiro[5.5]undecane dihydrochloride
2,9-Diazaspiro[5.5]undecane dihydrochloride
2,9-Diazaspiro[5.5]undecane dihydrochloride
APOSOR304403-1G
:
Apollo Scientific
:
2,9-Diazaspiro[5.5]undecane dihydrochloride
APOSOR304403-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
227.175 g/mol
:
Ambient
:
2227206-72-4
:
C₉H₁₈N₂·2ClH
:
MFCD10700111