7-Amino-2-azaspiro[4.4]nonan-3-one hydrochloride
Catalog # APOSOR304401-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:190.671 g/mol
Storage Temperature:Ambient
CAS Number:2173991-79-0
Formula:C₈H₁₄N₂O·ClH
MDL Number:MFCD31617948