Order Entry
Export
ContactUsLinkComponent
 
3-Ethoxy-2,2-dimethylcyclobutan-1-amine
Catalog # APOSOR304381-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Ethoxy-2,2-dimethylcyclobutan-1-amine
Catalog # APOSOR304381-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :143.227 g/mol
Storage Temperature:Ambient
CAS Number:1333759-16-2
Formula:C₈H₁₇NO
MDL Number:MFCD19982396