2,2-Dimethylcyclobutan-1-ol
Catalog # APOSOR304366-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:100.159 g/mol
Storage Temperature:Ambient
CAS Number:35301-44-1
Formula:C₆H₁₂O
MDL Number:MFCD26959233