2,2-Dimethylcyclobutan-1-one
Catalog # APOSOR304354-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:98.143 g/mol
Storage Temperature:Ambient
CAS Number:1192-14-9
Formula:C₆H₁₀O
MDL Number:MFCD22049843