Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,3-Dimethylcyclobutan-1-amine hydrochloride
3,3-Dimethylcyclobutan-1-amine hydrochloride
3,3-Dimethylcyclobutan-1-amine hydrochloride
3,3-Dimethylcyclobutan-1-amine hydrochloride
APOSOR304345-1G
:
Apollo Scientific
:
3,3-Dimethylcyclobutan-1-amine hydrochloride
APOSOR304345-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
135.635 g/mol
:
Ambient
:
1284247-23-9
:
C₆H₁₃N·ClH
:
MFCD22987614