(2S)-3,3-Dimethylazetidine-2-carboxylic acid
Catalog # APOSOR304331-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:129.157 g/mol
Storage Temperature:Ambient
CAS Number:1860033-50-6
Formula:C₆H₁₁NO₂
MDL Number:MFCD30146439