3,6-Dichloro-4-hydroxypyridazine 95+%
Catalog # APOSOR304303-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:164.977 g/mol
Storage Temperature:Ambient
CAS Number:2779-81-9
Formula:C₄H₂Cl₂N₂O
MDL Number:MFCD04971276