Order Entry
Export
ContactUsLinkComponent
1,3-Acetonedicarboxylic acid anhydride 97%
1,3-Acetonedicarboxylic acid anhydride 97%
Catalog # APOSOR30430-250MG
Supplier:  Apollo Scientific
CAS Number:  
1,3-Acetonedicarboxylic acid anhydride 97%
Catalog # APOSOR30430-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :128.083 g/mol
Storage Temperature:Ambient
CAS Number:10521-08-1
Formula:C₅H₄O₄
MDL Number:MFCD07783094