Order Entry
Export
Orders LinkContactUsLinkComponent
3,4-Dinitrobenzaldehyde

3,4-Dinitrobenzaldehyde

Irritant
New Product
Supplier:   Apollo Scientific
3,4-Dinitrobenzaldehyde
Supplier:  Apollo Scientific
New Product
Certificates
3,4-Dinitrobenzaldehyde

3 Options Available Below

Product Details & Documents
  • CAS Number:35998-98-2
  • Formula:C₇H₄N₂O₅
  • MDL Number:MFCD00845054
  • Molecular Weight:196.117 g/mol
  • Melting Pt:83…85 °C
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 213,00 €
    213,00 €
    1 * 500 mg
       213,00 €

  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 335,00 €
    335,00 €
    1 * 1 g
       335,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 1 510,00 €
    1 510,00 €
    1 * 5 g
       1 510,00 €

    Specifications:
    New Product
    New Product