
1-Benzyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trimethylsilyl)-1H-1,2,3-triazole, tech
Catalog # APOSOR303324-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:357.33 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1130489-19-8
Formula:C₁₈H₂₈BN₃O₂Si
MDL Number:MFCD18383356