Order Entry
Export
ContactUsLinkComponent
(2E)-3-{1,4-Dioxaspiro[4.5]dec-7-en-7-yl}prop-2-enoic acid
(2E)-3-{1,4-Dioxaspiro[4.5]dec-7-en-7-yl}prop-2-enoic acid
Catalog # APOSOR303181-500MG
Supplier:  Apollo Scientific
CAS Number:  
(2E)-3-{1,4-Dioxaspiro[4.5]dec-7-en-7-yl}prop-2-enoic acid
Catalog # APOSOR303181-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :210.226 g/mol
Storage Temperature:Ambient
CAS Number:226916-19-4
Formula:C₁₁H₁₄O₄
MDL Number:MFCD13152280