Order Entry
Export
Orders LinkContactUsLinkComponent
 

Quinoline-4,7-diol

Supplier:   Apollo Scientific
Quinoline-4,7-diol
Supplier:  Apollo Scientific
Certificates
Quinoline-4,7-diol

5 Options Available Below

Product Details & Documents
  • CAS Number:955938-89-3
  • Formula:C₉H₇NO₂
  • MDL Number:MFCD09260461
  • Molecular Weight:161.157 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 21,80 €
    21,80 €
    1 * 250 mg
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 42,00 €
    42,00 €
    1 * 1 g
       42,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 203,00 €
    203,00 €
    1 * 5 g
       203,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 801,00 €
    801,00 €
    1 * 25 g
       801,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 3 160,00 €
    3 160,00 €
    1 * 100 g
       3 160,00 €

    Specifications:
    New Product
    New Product