Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl 4,6-dichloro-2-methylnicotinate
Ethyl 4,6-dichloro-2-methylnicotinate 97%
Ethyl 4,6-dichloro-2-methylnicotinate 97%
Irritant
Ethyl 4,6-dichloro-2-methylnicotinate 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Ethyl 4,6-dichloro-2-methylnicotinate 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
686279-09-4
:
Formula:
C₉H₉Cl₂NO₂
MDL Number:
MFCD05666193
:
234.079 g/mol
:
GHS07
Storage Temperature:
Ambient