Order Entry
Export
ContactUsLinkComponent
Pyrazolo[1,5-a]pyrimidine 98%
Pyrazolo[1,5-a]pyrimidine 98%
Catalog # APOSOR301222-1G
Supplier:  Apollo Scientific
CAS Number:  
Pyrazolo[1,5-a]pyrimidine 98%
Catalog # APOSOR301222-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :119.124 g/mol
Storage Temperature:Ambient
CAS Number:274-71-5
Formula:C₆H₅N₃
MDL Number:MFCD10699580