
5-Bromo-4,6-dimethyl-2-hydroxynicotinic acid
Catalog # APOSOR300592-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:246.058 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:339366-43-7
Formula:C₈H₈BrNO₃
MDL Number:MFCD19981490