Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-6-chloro-1H-pyrrolo[3,2-c]pyridine
3-Bromo-6-chloro-1H-pyrrolo[3,2-c]pyridine
3-Bromo-6-chloro-1H-pyrrolo[3,2-c]pyridine
Irritant
3-Bromo-6-chloro-1H-pyrrolo[3,2-c]pyridine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3-Bromo-6-chloro-1H-pyrrolo[3,2-c]pyridine
Supplier:
Apollo Scientific
About this item
CAS Number:
1000341-61-6
:
Formula:
C₇H₄BrClN₂
MDL Number:
MFCD09749982
:
231.477 g/mol
:
GHS07
Storage Temperature:
Ambient