Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
8-Chloro-6-iodoimidazo[1,2-a]pyrazine
8-Chloro-6-iodoimidazo[1,2-a]pyrazine
8-Chloro-6-iodoimidazo[1,2-a]pyrazine
Irritant
8-Chloro-6-iodoimidazo[1,2-a]pyrazine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
8-Chloro-6-iodoimidazo[1,2-a]pyrazine
Supplier:
Apollo Scientific
About this item
CAS Number:
2227272-49-1
:
Formula:
C₆H₃ClIN₃
MDL Number:
MFCD31561275
:
279.466 g/mol
:
GHS07
Storage Temperature:
Ambient