
6-Iodo-8-phenylimidazo[1,2-a]pyrazine
Catalog # APOSOR300224-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:321.116 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:2227272-86-6
Formula:C₁₂H₈IN₃
MDL Number:MFCD31561255