Order Entry
Export
ContactUsLinkComponent
6-Bromo-8-phenylimidazo[1,2-a]pyrazine
Irritant
6-Bromo-8-phenylimidazo[1,2-a]pyrazine
Catalog # APOSOR300223-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
6-Bromo-8-phenylimidazo[1,2-a]pyrazine
Catalog # APOSOR300223-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :274.116 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1159815-57-2
Formula:C₁₂H₈BrN₃
MDL Number:MFCD12033252