Order Entry
Export
ContactUsLinkComponent
6-Bromo-8-chloro-[1,2,4]triazolo[4,3-a]pyrazine
6-Bromo-8-chloro-[1,2,4]triazolo[4,3-a]pyrazine
Catalog # APOSOR300030-1G
Supplier:  Apollo Scientific
CAS Number:  
6-Bromo-8-chloro-[1,2,4]triazolo[4,3-a]pyrazine
Catalog # APOSOR300030-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :233.453 g/mol
Storage Temperature:Ambient
CAS Number:1334135-75-9
Formula:C₅H₂BrClN₄
MDL Number:MFCD20501878