
3-Bromo-5-chloroimidazo[1,2-a]pyrazine
Catalog # APOSOR300023-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:232.465 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1243389-43-6
Formula:C₆H₃BrClN₃
MDL Number:MFCD16995965