
Ethyl 4-methyl-2-phenyl-1,3-thiazole-5-carboxylate
Catalog # APOSOR29649-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:247.313 g/mol
Boiling Pt:150 °C (0.4 mm Hg)
Melting Pt:36…39 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:53715-64-3
Formula:C₁₃H₁₃NO₂S
MDL Number:MFCD00141953