ethyl 2-(1,4-dimethyl-3-oxo-2-piperazinyl)acetate
Catalog # APOSOR29248-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:214.262 g/mol
Storage Temperature:Ambient
CAS Number:175205-86-4
Formula:C₁₀H₁₈N₂O₃
MDL Number:MFCD00178772