2,5-Dimethoxythiophene 95+%
Catalog # APOSOR29056-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:144.192 g/mol
Storage Temperature:Ambient
CAS Number:58386-20-2
Formula:C₆H₈O₂S
MDL Number:MFCD01096545