
5-(4-Methylphenyl)-1,3-oxazole 97%
Catalog # APOSOR29030-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:159.185 g/mol
Melting Pt:65…68 °C
:GHS07
Storage Temperature:Ambient
CAS Number:143659-19-2
Formula:C₁₀H₉NO
MDL Number:MFCD00085134