
Ethyl 1-methylcyclopropane-1-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:71441-76-4
:
Formula:C₇H₁₂O₂
MDL Number:MFCD00001284
:128.169 g/mol
Boiling Pt:136 °C (760 mm Hg)
Density:0.918 g/cm³ (25 °C)
Flash Pt:30 °C
Storage Temperature:Refrigerator
EINECS:275-461-6
UN:3272
ADR:3,III
:GHS02