Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Bis(4-phenoxybenzoyl)benzene
1,4-Bis(4-phenoxybenzoyl)benzene
1,4-Bis(4-phenoxybenzoyl)benzene
APOSOR2850-1G
:
Apollo Scientific
:
1,4-Bis(4-phenoxybenzoyl)benzene
APOSOR2850-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
470.515 g/mol
:
216…218 °C
:
GHS07
:
Ambient
:
54299-17-1
:
C₃₂H₂₂O₄
:
MFCD00319174