Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Bromo-4,5-dimethyl-2,1,3-benzothiadiazole
6-Bromo-4,5-dimethyl-2,1,3-benzothiadiazole
6-Bromo-4,5-dimethyl-2,1,3-benzothiadiazole
APOSOR26817-1G
:
Apollo Scientific
:
6-Bromo-4,5-dimethyl-2,1,3-benzothiadiazole
APOSOR26817-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
243.124 g/mol
:
84…85 °C
:
GHS07
:
Ambient
:
175204-24-7
:
C₈H₇BrN₂S
:
MFCD00126470