
6-Methylimidazo[2,1-b][1,3]thiazole-5-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:77628-51-4
:
Formula:C₇H₆N₂O₂S
MDL Number:MFCD02083132
:182.2 g/mol
Melting Pt:164…165 °C
:GHS07
Storage Temperature:Ambient