2,4-Dichlorophenylhydrazine
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: (2,4-Dichlorophenyl)hydrazine
- CAS Number:13123-92-7
- Formula:C₆H₆Cl₂N₂
- MDL Number:MFCD00044514
- Molecular Weight:177.031 g/mol
- Melting Pt:91…94 °C
- Storage Temperature:Refrigerator
3 Options Available Below
Product Details & Documents
Synonyms: (2,4-Dichlorophenyl)hydrazine
- CAS Number:13123-92-7
- Formula:C₆H₆Cl₂N₂
- MDL Number:MFCD00044514
- Molecular Weight:177.031 g/mol
- Melting Pt:91…94 °C
- Storage Temperature:Refrigerator



