2,6-Dimethyldecane
Bottle, 1 g
Catalog # APOSOR2375-1G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
- CAS Number:13150-81-7
- Formula:C₁₂H₂₆
- MDL Number:MFCD00060964
- Molecular Weight:170.335 g/mol
- Storage Temperature:Ambient
- UN:1993
- ADR:3,III
Specifications
Specifications
- Pack type:Bottle
- Pk:1 g



