Order Entry
Export
ContactUsLinkComponent
4-(3-Bromophenyl)-2-methyl-1,3-thiazole 95%
Irritant
4-(3-Bromophenyl)-2-methyl-1,3-thiazole 95%
Catalog # APOSOR23314-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-(3-Bromophenyl)-2-methyl-1,3-thiazole 95%
Catalog # APOSOR23314-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:56…65 °C
 :254.146 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:342405-21-4
Formula:C₁₀H₈BrNS
MDL Number:MFCD02681947