Order Entry
Export
ContactUsLinkComponent
Ethyl 2,4-dichlorophenylacetate 97%
Irritant
Ethyl 2,4-dichlorophenylacetate 97%
Catalog # APOSOR2296-500MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl 2,4-dichlorophenylacetate 97%
Catalog # APOSOR2296-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :233.091 g/mol
Boiling Pt:88 °C (0.05 mm Hg)
 :GHS07
Storage Temperature:Ambient
CAS Number:41022-54-2
Formula:C₁₀H₁₀Cl₂O₂
MDL Number:MFCD01566538