
2,6-Dibromonitrobenzene 95+%
Catalog # APOSOR2213-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:280.902 g/mol
Melting Pt:78…82 °C
:GHS07
Storage Temperature:Ambient
CAS Number:13402-32-9
Formula:C₆H₃Br₂NO₂
MDL Number:MFCD09998726