
3-Aminothieno[2,3-c]pyridine
Catalog # APOSOR2143-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:150.201 g/mol
Melting Pt:148…150 °C
:GHS07
Storage Temperature:Ambient
CAS Number:63326-75-0
Formula:C₇H₆N₂S
MDL Number:MFCD09998716