

3-Allylphenol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1446-24-8
:
Formula:C₉H₁₀O
MDL Number:MFCD20257851
:134.175 g/mol
Boiling Pt:108…115 °C (13 mm Hg)
UN:2922
ADR:8+6.1,III
:GHS05
:GHS06
Storage Temperature:Ambient