2,6-Dichloro-4-methylbenzaldehyde 95+%
Catalog # APOSOR17621-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:189.039 g/mol
Storage Temperature:Ambient
CAS Number:116070-31-6
Formula:C₈H₆Cl₂O
MDL Number:MFCD09753749