0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
2-Methyl-1,3-oxazole-4-carbohydrazide
2-Methyl-1,3-oxazole-4-carbohydrazide
2-Methyl-1,3-oxazole-4-carbohydrazide
2-Methyl-1,3-oxazole-4-carbohydrazide
APOSOR17536-250MG
:
Apollo Scientific
:
2-Methyl-1,3-oxazole-4-carbohydrazide
APOSOR17536-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
141.128 g/mol
:
GHS07
:
Refrigerator
:
500341-65-1
:
C₅H₇N₃O₂
:
MFCD11052651