Order Entry
Export
ContactUsLinkComponent
3,7-Dihydro-5-iodo-4H-pyrrolo[2,3-d]pyrimidin-4-one
Irritant
3,7-Dihydro-5-iodo-4H-pyrrolo[2,3-d]pyrimidin-4-one
Catalog # APOSOR17053-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3,7-Dihydro-5-iodo-4H-pyrrolo[2,3-d]pyrimidin-4-one
Catalog # APOSOR17053-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :261.02 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:135352-71-5
Formula:C₆H₄IN₃O
MDL Number:MFCD09746273