0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
7-Oxa-1-azaspiro[4.4]nonane hydrochloride
7-Oxa-1-azaspiro[4.4]nonane hydrochloride
7-Oxa-1-azaspiro[4.4]nonane hydrochloride
:
Apollo Scientific
7-Oxa-1-azaspiro[4.4]nonane hydrochloride
:
Apollo Scientific
:
1620569-18-7
:
:
C₇H₁₃NO·ClH
:
MFCD26960527
:
163.645 g/mol
:
GHS07
:
Ambient